ContactUsLinkComponent
Glycogen Assay Kit, Cayman Chemical Company
Glycogen Assay Kit, Cayman Chemical Company
# de catalogue 75817-144
Numéro CAS:  
KIT GLYCOGEN ASSAY 2.5-40 UG/ML 96 WELLS
Glycogen Assay Kit, Cayman Chemical Company
# de catalogue 75817-144
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75817-144
  • Contents:
    Kit contains: Standard, Buffers, Enzyme, Enzyme Mixture, Detector, and Plate
  • Supplier No.:
    700480-96
Spécifications
A propos de cet article
A simple and sensitive tool for assaying glycogen from tissue
  • Measure glycogen from tissue samples
  • Assay 20 samples in duplicate
  • Assay Range: 2.5-40 µg/ml
  • Plate-based fluorometric measurement (ex 530-540 nm, em 585-595 nm)
Glycogen is the principal storage form of glucose in animal and human cells. It is made primarily by the liver and muscles, but it can also be made by glycogenesis within the brain and stomach. Cayman’s Glycogen Assay provides a simple, reproducible, and sensitive tool for assaying glycogen from tissue. In this assay, glycogen is hydrolyzed by amyloglucosidase to form β-D-glucose, which is then specifically oxidized to D-glucono-δ-lactone by glucose oxidase forming hydrogen peroxide in the process. Hydrogen peroxide, in the presence of horseradish peroxidase, reacts with 10-acetyl-3,7-dihydroxyphenoxazine (ADHP) in a 1:1 stoichiometry to generate the highly fluorescent product resorufin which is measure at an excitation wavelength of 530-540 nm and an emission wavelength of 585-595 nm.
Customers Who Bought This Also Bought