A propos de cet article
For the measurement of catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates
- Measure catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, and cell lysates
- Assay 40 samples in duplicate
- Measure catalase activity down to 2 U/ml
- Plate-based colorimetric measurement (540 nm)
Catalase is an ubiquitous antioxidant enzyme that is present in most aerobic cells. It is involved in the detoxification of H2O2, a ROS that is a toxic product of both normal aerobic metabolism and pathogenic ROS production. Cayman’s Catalase Assay utilizes the peroxidatic function of catalase for determination of enzyme activity.