Spécifications
- Description:BA410E Premium Phase Lab Package
- Condenser:5 position (BF, DF, PH1, PH2, PH3) turret condenser
- Eyepieces:Widefield high eyepoint eyepiece WFPL 10×/22 mm with diopter adjustment and rubber eyecup
- Focusing system:Coaxial Coarse and Fine Knobs
- Head:Siedentopf, 30° Inclined, 360° Rotating, 20:80 Light Split
- Illumination:12V/50W Koehler Halogen / 12V3W LED Module (6000K)
- Nosepiece:Sextuple, Reversed
- Objectives:CCIS plan achromatic EC-H PL 4× and CCIS plan phase EC-H Ph 10×, EC-H Ph 40× S, EC-H Ph 100× S-oil
- Cat. no.:10854-394
- Supplier No.:1100100403242
Spécifications
A propos de cet article
Motic's BA410 Elite Phase Package is a complete, premium phase microscopy bundle. This bundle is composed of the highest quality Motic components as well as a wide range of accessories and upgrades making it the perfect choice for any professional life-science research microscopist with a need for phase contrast microscopy.
- Improved interpupillary distance and moveable eyepieces result in maximum comfort for any user
- Rackless stage can hold two slides at once for quick comparison
- Absence of gear rack on the stage allows for quick throughput of slides
- High-end CCIS Achromatic optics minimize chromatic and spherical aberrations
- A variety of optional upgrades and accessories is available
- A phase turret condenser offers maximum flexibility with all necessary illumination rings
The BA410 Elite Phase Package is Motic's premium, professional option for phase contrast microscopy.
The BA410 Elite's CCIS Plan Achromat Objectives are designed for maximum image fidelity. The expanded resolution power in terms of numerical aperture, along with impeccable color reproduction, yield the most accurate sample representation.
Attention has also been paid to ergonomics and comfort. With adjustable eyepieces and rubber eyecups, this microscope can be used for extended periods of time without strain on the eyes or neck.
With a trinocular head, this microscope comes ready for photomicrography. Each part of the BA410 Elite has been designed with the understanding of the importance of reproducible documentation. Just add a camera and appropriate c-mount adapter, and you will have a system ready to deliver the highest quality digital images.