Spécifications
- Description:Motic BA410E Upright Compound Microscope with Complete Set of Cytology Accessories
- Condenser:Achromat Swing-out Condenser N.A.0.90 W/ Iris Diaphragm
- Eyepieces:Widefield high eyepoint eyepiece WFPL 10×/22 mm with diopter adjustment
- Head:Siedentopf, 30° Inclined
- Illumination:Koehler Illumination Quartz Halogen 12V/50W and 12V/3W led module with external lamp house and intensity control
- Nosepiece:Sextuple, Reversed
- Objectives:CCIS EC Plan Achromatic EC PL 4×, 10×, 40× S, 60× S
- Stage:Built in low position rackless stage with right hand control
- Cat. no.:10854-400
- Supplier No.:1100100403049
Spécifications
A propos de cet article
The BA410 Elite Cytology Package is a complete cytology microscope bundle, which includes the Motic's flagship life-science microscope, the BA410 Elite. This package has been crafted to provide everything needed to perform professional level cytology microscopy with efficiency and effectiveness.
- Improved interpupillary distance and moveable eyepieces result in maximum comfort for any user
- Rackless stage can hold two slides at once for quick comparison
- Absence of gear rack on the stage allows for quick throughput of slides
- High-end CCIS Achromatic optics minimize chromatic and spherical aberrations
- A variety of optional upgrades and accessories is available
The BA410 Elite Cytology Package comes complete with all the accessories needed for cytology microscope applications.
The BA410 Elite's CCIS Plan Achromat Objectives are designed for maximum image fidelity. The expanded resolution power in terms of numerical aperture, along with impeccable color reproduction, yield the most accurate sample representation.
Attention has also been paid to ergonomics and comfort. With adjustable eyepieces and rubber eyecups, this microscope can be used for extended periods of time without strain on the eyes or neck.
With a trinocular head, this microscope comes ready for photomicrography. Each part of the BA410 Elite has been designed with the understanding of the importance of reproducible documentation. Just add a camera and appropriate c-mount adapter and you will have a system ready to deliver the highest quality digital images.