Cool Offs, Allegro
Fournisseur: Allegro
Certificats
A propos de cet article
Cooling products are designed to protect and provide comfort to workers who must function in environments with excessive heat.
- Lightweight and comfortable
- 38" length (longest on the market)
- 100% cotton
Keep cool all day long with these dual-use Cool Offs
To use, simply soak the garment in cold water for 10 to 15 minutes. The cooling crystals go to work to keep wearers cool for days.
Quilted into four sections for an even distribution of hydration material, Cool Offs can be worn around the neck or tied as a headband. A unique button hole closure ensures a secure fit. The outer shell is 100% cotton and comes in multiple colours and patterns.
Spécifications
- Catalog No:
- 76169-212
- 76169-218
- 76169-220
- 76169-214
- 76169-216
- 76169-222
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton
- 100% cotton