VWR® Key Pad and Keyless Access Locks for Refrigerators and Freezers
Fournisseur: VWR International
Certificats
A propos de cet article
Key Pad Access and Keyless Access locks for various refrigerators and freezers.
- Durable
- Battery operated
- Programmable passcodes
Key Pad Access Locks for 12, 23 and 26 cu. ft. VWR® Performance, VWR® Series and VWR® Standard Upright Solid Door Refrigerators, Undercounter Built-In (UCBI) Refrigerators and Freezers, and Undercounter Freestanding (UCFS) Refrigerators and Freezers (excluding the General Purpose models, 10819-650 and 10819-894).
Keyless Access lock for Undercounter built-in (UCBI) refrigerator and freezer models.
Keyless Access lock for Undercounter freestanding (UCFS) refrigerator and freezer models.
Customers Who Bought This Also Bought