dNTP Mix, Zymo Research
Fournisseur: Zymo Research
Certificats
A propos de cet article
The dNTP Mix, along with dATP, dTTP, dGTP, and dCTP mixes from Zymo Research are of ultra-high purity and can be used to generate DNA by PCR using ZymoTaq™ or other DNA polymerases.
- Ready to use dNTP Mix (dATP, dTTP, dGTP, dCTP) of ultra high purity; >99% trisphosphate by HPLC
- Readily incorporated into PCR amplicons with ZymoTaq™ or other DNA polymerases
- Free of endo-, exodeoxyribonuclease, ribonuclease, phosphatase, and nicking activities
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.