ContactUsLinkComponent
EISCO® Ebola Viron Model
EISCO® Ebola Viron Model
# de catalogue 470231-596
Fournisseur:  Wards
Model Ebola Virus 10000X
EISCO® Ebola Viron Model
# de catalogue 470231-596
Fournisseur:  Wards
Spécifications
  • Description:
    Ebola Model
  • Cat. no.:
    470231-596
Spécifications
A propos de cet article
The Only Ebola Model Available!
  • Approximately 100000X
  • Includes keycard
First described in 1976, the Ebola virus in one of the most deadly viruses on Earth. Once infected, patients develop severe hemorrhagic fever, which has a 90% mortality rate. There have been several outbreaks in Zaire, Sudan, Congo and Uganda, and also a few cases of fatalities due to laboratory accidents. Ebola is one of the few virus that needs to be contained in a Level 4 Biohazard facility.
This model represents the virus magnified approximately 100,000 times in its unique shape. The surface of the model shows Glycoproteins viral membrane. A cut- away at one end exposes internal structures, major and minor matrix proteins, polymerase protein, and Ebola RNA. Included key card.
Customers Who Bought This Also Bought