Assay Plates, 96-Well, Thomson Instrument
Fournisseur: THOMSON INSTRUMENT CO
New Product
Certificats
A propos de cet article
These 96-well assay plates are used for most common range of working volumes.
- Made of LCMS tested PP
For long term storage use cap mat over a heat seal, but researchers have to be careful when defrosting. It is best to remove the capmat and replace with heat seal to avoid condensation raining into wells while thawing.
Customers Who Bought This Also Bought