SpectroMicro® Sample Cups, Chemplex
Fournisseur: Chemplex
Certificats
A propos de cet article
Vented Snap-On Cap equalizes pressure differentials within the Cell and sample chamber to maintain a taut thin-film sample support plane.
- Larger conically shaped upper portion of the funnel allows and facilitates sample introduction
- Lower cylindrically shaped spout of the funnel represents the actual sample chamber
The collar is used to secure a thin-film sample support window substance to the Cell and designed for acceptance by 32 mm sample cup holders.
Customers Who Bought This Also Bought