AIRFREE® Filter Tubes, Schlenk, Chemglass
These AIRFREE® filter tube allows for filtration followed by collection of crystals on opposite side of fritted disc.
- Standard taper inner joints at both ends are the same size
- Available with a choice of coarse or medium porosity fritted discs
Customers Who Bought This Also Bought