Nomex® Flame-Resistant Lab Coat, Men's/Unisex , CritiCore Protective Wear
Fournisseur: VWR International
Certificats
A propos de cet article
Designed to provide a superior level of protection against flash fire, electric arc (HRC 1), and other dangerous conditions, this Flame-Resistant (FR) lab coat has numerous applications in both laboratory and production settings. Made with Nomex® CXP6 fabric, it combines exceptional flame resistance with excellent durability, even after repeated industrial launderings.
- Excellent heat-resistance properties
- Best in class flammability protection
- 360 degree FR identification and protection
- Meets NFPA 70E and HRC 1 performance standards
- Quick-release snap front
- Safety sleeve cuff with tunneled elastic
Coats feature 360 degree FR identification with patches sewn on the back, plus left and right sleeves
Nomex® is a registered trademark owned by E. I. du Pont de Nemours and Company.
Constructed with a quick-release snap front and safety sleeve cuff with tunneled elastic for additional protection. Coats also feature two lower hip pockets and one breast pocket with pencil slot.
Certifications: Meets performance standards for NFPA 2112 and NFPA 70E HRC 1.
Spécifications
- Catalog No:
- 76164-544
- Garment application:
- Industrial, Electronics/semi-conductor, Laboratory
- Risk area:
- Anti-static, Flame-retardant
- Gender:
- Unisex
- Sleeve or leg length:
- Long
- Closure type:
- Press studs / snaps
- Fabric:
- Nomex® CXP6