Multi-Purpose Containers with Snap Cap, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
These disposable containers are produced from high density polyethylene and are ideal for the collection and storage of histology and pathology specimens, fluids, powders, and solids.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.