Rapid Clear® Cap 3000, Thomson Instrument Company
# de catalogue 76808-350
Fournisseur: THOMSON INSTRUMENT CO
Spécifications
- Sterility:Sterile
- Description:Rapid Clear® cap 3000
- Cat. no.:76808-350
Spécifications
A propos de cet article
Designed for high-speed clarification of 2 to 4 litres of cell culture.
- Save time by clarifying 3 litres of cell culture in less than 35 minutes
- 0.2 μm filter eliminates multiple filtration steps, and in some cases centrifugation
- Cell clarification of low or high density cultures of CHO stable, CHO transient, HEK293, hybridoma, and other mammalian cell lines
- Eliminate transfer steps: The Rapid Clear® Cap screws directly onto Optimum Growth™ Flasks
Use with 2.8 and 5 litres Optimum Growth™ Flasks.
Customers Who Bought This Also Bought