Screw Thread Culture Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
These borosilicate culture tubes are suitable for tissue culture work and general bacteriological use.
- Round bottom and supplied without caps
- No marking spot
Modular trays are shrink-wrapped in plastic film.
Customers Who Bought This Also Bought