About this item
Huber Immersion Coolers are easy to use and ideally suited to quick liquid cooling.
- Universal solution for many cooling tasks
- Rapid cooling of fluids
- Ideal for counter-cooling or as a replacement of dry ice
- LED temperature display
- Easy operation with only three keys
The TC range of Immersion Coolers are a flexible solution for a variety of cooling tasks
The models with the type affix "E“ have an LED display with entry of nominal values and regulate with a precision of ±0.5 K. A spiral-shaped or flexible immersion cooling probe made out of stainless steel is available for all models so that fluids can be cooled in virtually all vessel types. The compact housing is made from high quality stainless steel, and is fitted with carrying grips. Their small footprints require little installation area and roll castors are available as an accessory for simple changes of location.
All models can be delivered with a flexible evaporator coil (no extra cost). The model name and number of these units have the suffix ”F“.
The devices are simple and universally used for rapid cooling of fluids. Typical applications include counter-cooling for heat circulation thermostats or replacement of dry ice. For many applications Immersion Coolers are also an environmentally friendly alternative to mains water cooling. The TC Immersion Coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a Pt100 probe.
Specifications
- Catalog No:
- 10221-076
- 10221-070
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Cooling capacity:
- 0.12 kW ( at -80 °C); 0.16 kW (at 0 °C)
- 0.12 kW ( at -80 °C); 0.16 kW (at 0 °C)