Spécifications
- Description:Lipid Test Strips
- Cat. no.:470093-682
- Supplier No.:LIP-1B-75
- No. of Tests:75
Spécifications
A propos de cet article
A simple dip test for presence of lipids in solution.
- Resealable zip closure bag
- Waterproof chart
Easily test solutions for the presence of fats with these single use dip tests. Each strip had a single testing pad which will change color in the presence of lipids. Simply dip, wait 2 seconds and compare to the chart.