Protein Test Strips
# de catalogue 470092-374
Fournisseur: Precision Laboratories
Spécifications
- Description:Protein Test Strips
- Cat. no.:470092-374
- Supplier No.:PRO-1V-50
- No. of Tests:50
Spécifications
A propos de cet article
Add protein testing to your urinalysis procedures.
- Flip top vial
- Waterproof color chart
These single-test test strips have a plastic strip with a reactive pad to prevent contact contamination while handling. Simply dip the pad into the solution, wait 60 seconds, and compare to the color chart on the side of the bottle.
Caution: Flip top vial
Waterproof color chart
Customers Who Bought This Also Bought