ContactUsLinkComponent
VWR® Beakers without Handle, PP
VWR® Beakers without Handle, PP
Catalog # 76019-260
Supplier:  VWR International
   Sustainable Options Available
Sustainable Packaging
VWR® Beakers without Handle, PP
VWR® Beakers without Handle, PP
Catalog # 76019-260
Supplier:  VWR International
   Sustainable Options Available
  Sustainable Packaging
Log in for availability
Select an Option
42.5 mm
Select an Option
5 ml
Select an Option
5 - 25 ml
Select an Option
25 ml (0.85 oz.)
Select an Option
49.8 mm
Select an Option
76019-260
Specifications
  • Catalog No:
  • 76019-260
  • 76019-262
  • 76019-264
  • 76019-266
  • 76019-268
  • 76019-270
  • 76019-272
  • 76019-274
  • 76019-276
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
Specifications
About this item
VWR® polypropylene beakers, which come in a natural color, have a pouring spout and graduations printed directly on the beakers.
  • Autoclavable
  • Integrated pouring lip
  • Stackable
  • Natural color
  • Printed graduation
Since the beakers have no handle, they can be easily stacked to save space. The beakers are autoclavable.
Working temperatures of 0 to 121 °C.
Certifications: Resin meets US FDA 21 CFR requirements.
Caution: Do not stack products while autoclaving.
Customers Who Bought This Also Bought