VWR® TPX™ Beakers with Handle
À propos de cet article
VWR® TPX™ Beakers, which are crystal-clear in color, have a handle, pouring spout and graduations printed directly on the beakers.
- Chemical resistant
- Integrated pouring lip
- Autoclavable
- Natural color
- Printed graduation
The beakers are autoclavable and chemical resistant.
Working temperatures of 0 to 121 °C (32 to 248 °F) when used continuously.
Working temperatures as high as 170 °C (338 °F) when used for short periods of time.
: Resin meets US FDA 21 CFR requirements.
: Do not stack products while autoclaving.
6 Options Available Below
Product Details & Documents
VWR® TPX™ Beakers, which are crystal-clear in color, have a handle, pouring spout and graduations printed directly on the beakers.
- Chemical resistant
- Integrated pouring lip
- Autoclavable
- Natural color
- Printed graduation
The beakers are autoclavable and chemical resistant.
Working temperatures of 0 to 121 °C (32 to 248 °F) when used continuously.
Working temperatures as high as 170 °C (338 °F) when used for short periods of time.
: Resin meets US FDA 21 CFR requirements.
: Do not stack products while autoclaving.
Certificates
Recherche par numéro de lotSpecifications for Product Family
Spécifications
- Catalog No:
- 76019-278
- 76019-280
- 76019-282
- 76019-284
- 76019-286
- 76019-288
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Spécifications
Product Family Options
Informations sur le produit
- CapacitéHauteurØ ext.Intervalle de graduationPlage de graduationAvailabilityPrice
- 100 ml (3.4 oz.)73.5 mm63.4 mm20 ml20 - 100 mlLog in pour la disponibilitéCase of 12Case of 12 - $50.91$50.91Prix promotionnel pour les nouveaux clientsEach$4.24Case of 12$50.91Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-278Environmentally PreferableSustainable PackagingSustainable Packaging - 250 ml93 mm84 mm50 ml50 - 250 mlLog in pour la disponibilitéCase of 12Case of 12 - $93.28$93.28Prix promotionnel pour les nouveaux clientsEach$7.77Case of 12$93.28Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-280Environmentally PreferableSustainable PackagingSustainable Packaging - 500 ml128 mm110.5 mm100 ml100 - 500 mlLog in pour la disponibilitéCase of 12Case of 12 - $175.63$175.63Prix promotionnel pour les nouveaux clientsEach$14.64Case of 12$175.63Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-282Environmentally PreferableSustainable PackagingSustainable Packaging - 1000 ml150 mm129.4 mm100 ml100 - 1000 mlLog in pour la disponibilitéCase of 4Case of 4 - $117.58$117.58Prix promotionnel pour les nouveaux clientsEach$29.40Case of 4$117.58Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-284Environmentally PreferableSustainable PackagingSustainable Packaging - 2000 ml191 mm158 mm200 ml200 - 2000 mlLog in pour la disponibilitéCase of 4Case of 4 - $184.32$184.32Prix promotionnel pour les nouveaux clientsEach$46.08Case of 4$184.32Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-286Environmentally PreferableSustainable PackagingSustainable Packaging - 5000 ml262 mm203 mm1000 ml1000 - 5000 mlLog in pour la disponibilitéCase of 4Case of 4 - $322.45$322.45Prix promotionnel pour les nouveaux clientsEach$80.61Case of 4$322.45Spécifications:
Sustainable Packaging More
Specifications
Cat. No.76019-288Environmentally PreferableSustainable PackagingSustainable Packaging