BioFit Urethane Lab Chairs and Stools
Supplier: VWR International
Certificates
About this item
Lab Chairs and stools are made of urethane, crafted with user comfort, health and wellbeing in mind.
- Easy-to-clean seats and backrests that resist water, chemicals, paints and more
- Easily adjust seat-height from seated position
- Height and tilt adjustable backrests with integrated lumbar support
- Dual-wheel resistance casters enhance user safety by inhibiting seating from rolling away on hard floors or 2" high polyamide glides
- Adjustable and fixed height footrings
BioFit seating solutions are crafted with user comfort, health and wellbeing in mind
Provide LEED® compliant seating for healthcare, technology, education, lab, industrial, office and custom applications.
Products meet or exceed rigorous industry standard and carry a 13-year warranty with a lifetime warranty on the pneumatic mechanisms.
Specifications
- Catalog No:
- 75821-338
- 75821-340
- 75821-342
- 75821-344
- 75821-346
- 75821-348
- 75821-350
- 75821-352
- 75821-332
- 75821-354
- 75821-334
- 75821-336
- Model:
- 75821-338
- 75821-340
- 75821-342
- 75821-344
- 75821-346
- 75821-348
- 75821-350
- 75821-352
- 75821-332
- 75821-354
- 75821-334
- 75821-336
- For use with:
- Low benches/desks
- Medium height benches
- Medium height benches
- Medium height benches
- High benches
- High benches
- Low benches/desks
- Medium height benches
- High benches
- Medium height benches
- High benches
- Low benches/desks
- Frame colour/finish:
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Chrome
- Frame material:
- 25.6" Ø high profile cast aluminum
- 25.6" Ø high profile cast aluminum
- 25.6" Ø high profile cast aluminum
- 25.6" Ø high profile cast aluminum
- 23" Ø tubular steel
- 23" Ø tubular steel
- 23" Ø tubular steel
- 23" Ø tubular steel
- 25.6" Ø high profile cast aluminum
- 23" Ø tubular steel
- 25.6" Ø high profile cast aluminum
- 25.6" Ø high profile cast aluminum
- Seat/back color:
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Black
- Seat/back material:
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Self-skinned urethane
- Seat dimensions:
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- W18.5×D17×T1.5"
- Backrest dimensions:
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"
- W16.5×H12.5"