A propos de cet article
Cell-Mate3D µGel 40™ provides your cells with an ideal microenvironment for disease modeling and regenerative medicine research
- Simple and easy setup at room temperature
- Tissue-like stiffness better mimics in vivo extracellular environment
- Allows for emergence of complex microtissues/organoids
- Chemically Defined and Quantifiable Tool
- Replace up to 20%* of hydration fluid with other ECM components for further customization
It is ideal for cancer cells to model the tumor microenvironment and for stem cells to differentiate toward Central Nervous System tissue. The unique microgel design takes advantage of its chemically defined, tissue-like, biomimetic 3D technology. Thus, allowing for use as a quantitative tool with fewer cells (lower limit 12K) embedded within the 3D matrix. Moreover, the simplicity and ease of use offers a fast set-up in as few as 5 minutes unlike other 3D products in the market.