Ergonomic Drum Handler Power Lift, Model DM-1100- PL, Wesco
# de catalogue 89412-256
Fournisseur: Wesco
Spécifications
- Description:Ergonomic Drum Handler Power Lift
- Cat. no.:89412-256
- Weight:531 lbs.
- Supplier No.:240156
Spécifications
A propos de cet article
Ergonomic drum handler power lift handles most 30, 55 or 85 gallon drums made of steel, fiber and poly.
- Works with most drums with locking rings
- Up/Down remote on power units
- Power drive travel speed 120/min
- Lifts, transports, weighs and places drums on spill or standard pallets
Features spring-loaded jaws hold any 3/16" or higher drum rim, Two wheels and two swivel casters, 5×2" phenolic, and floor lock. Exclusive auto grip lock will not drop a drum if operated properly.
Customers Who Bought This Also Bought