Corning® Screw Capped Micro Tubes, Corning
Catalog # 75831-988
Supplier: Corning
Specifications
- Sterility:Sterile
- Volume:1.5 ml
- Color:Amber
- Description:Self Standing Microtubes, with Brown Screw Cap and O-rings
- Cat. no.:75831-988
- Supplier No.:SCT-150-SS-X-S
Specifications
About this item
Screw cap microtubes serve a broad range of laboratory needs including collection, processing, centrifugation, long-term storage and packaging/shipping of restriction enzymes and other reagents.
- Withstand up to 20000 RCF
- Certified RNase-, DNase- and endotoxin-safe for total sample integrity
Color coded caps provide a syringe port which is easily pierced for radioisotope labelling and fraction collecting.
Frequently Bought Together