Cleanert® IC Sample Clean-Up Cartridges for Ion Chromatography, Bonna-Agela Technologies
Fournisseur: Agela Technologies
Certificats
A propos de cet article
Cleanert IC series are used for removing matrix interferences such as phenolics, metalions, cations, anions, or hydrophobic substances encountered in many ion chromatography applications.
- IC cartridges are designed as luer inlet and outlet for easy operation
They can absorb metal ions and reduce interferential ion in the sample, thus prolong the lifetimes of the analytical columns. They also can be used for some trace-level determinations.
They also can connect several cartridges together for complex matrix. The sample solution can be passed through the IC cartridge by syringe, and also by vacuum manifold connected with an empty SPE cartridge as funnel.
Spécifications
- Catalog No:
- 89213-618
- 89213-596
- 89213-620
- 89213-622
- 89213-600
- 89213-602
- 89213-610
- 89213-588
- 89213-612
- 89213-590
- 89213-592
- 89213-614
- 89213-616
- 89213-594
- 89213-624
- 89213-604
- 89213-626
- 89213-606
- 89213-584
- 89213-608
- 89213-586
- Application:
- To remove transition metals and matrix elimination of alkali and alkaline earth metals
- To remove anion contaminant and neutralize the strongly acidic sample solution
- Equivalent function as combined Ag and H type cartridges
- Equivalent function as combined Ag and H type cartridges
- To remove high levels of alkaline earths and transition metals from sample matrices and neutralization of highly alkaline samples
- To remove high levels of alkaline earths and transition metals from sample matrices and neutralization of highly alkaline samples
- To remove chloride, bromide and iodide from sample matrices
- To remove substances such as aromatic dyes, carboxylic acids, hydrocarbons and surfactants from sample matrices
- To remove SO42-, catridge should be activated with Cl- solution if the anion concentration is low
- To remove substances such as aromatic dyes, carboxylic acids, hydrocarbons and surfactants from sample matrices
- To remove phenolic fraction of humic acids, tannic acids, lignins, anthocyanins and azodyes from samples
- To remove SO42-, catridge should be activated with Cl- solution if the anion concentration is low
- To remove transition metals and matrix elimination of alkali and alkaline earth metals
- To remove phenolic fraction of humic acids, tannic acids, lignins, anthocyanins and azodyes from samples
- Equivalent function as combined Ag and Na type cartridges
- To remove high levels of alkaline earths and transition metals from sample matrices without acidifying the sample
- Equivalent function as combined Ag and Na type cartridges
- To remove high levels of alkaline earths and transition metals from sample matrices without acidifying the sample
- To remove the hydrophobe from biological samples
- To remove chloride, bromide and iodide from sample matrices
- To remove the hydrophobe from biological samples
- Exchange capacity:
- 0.4 meq/1 cc
- 0.7 meq/1 cc
- —
- —
- 2.0-2.2 meq/1 cc
- 2.0-2.2 meq/1 cc
- 2.0-2.2 meq/1 cc
- 300 mg/1 cc
- 2.0-2.2 meq/1 cc
- 300 mg/1 cc
- 350 mg/1 cc
- 2.0-2.2 meq/1 cc
- 0.4 meq/1 cc
- 350 mg/1 cc
- —
- 2.0-2.2 meq/1 cc
- —
- 2.0-2.2 meq/1 cc
- —
- 2.0-2.2 meq/1 cc
- —