ContactUsLinkComponent
EZ-Vac™ Vacuum Manifold, Zymo Research
EZ-Vac™ Vacuum Manifold, Zymo Research
Catalog # 76005-132
Supplier:  Zymo Research
EZ-Vac* Vacuum Manifold LLDPE manifold body
EZ-Vac™ Vacuum Manifold, Zymo Research
Catalog # 76005-132
Supplier:  Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EZ-Vac™ Vacuum Manifold
  • Cat. no.:
    76005-132
Specifications
About this item
The EZ-Vac™ Vacuum Manifold features durable chemical-resistant construction and is capable of processing up to 20 samples simultaneously using vacuum pressure without the need for repeated centrifugation steps
  • Compatible with many spin-column based nucleic acid purification kits
  • Unique design accommodates large processing volumes easily
  • Simplifies the nucleic acid purification workflows
  • The One-Way Luer-lock Stopcocks are individually controlled, allowing users to process as little as one sample at a time
The EZ-Vac™ Vacuum Manifold consist of a linear, low-density polyethylene (LLDPE) manifold body, One-Way Luer-Lock Stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector and a neoprene stopper.
Customers Who Bought This Also Bought