Heratherm™ Advanced Protocol Security Microbiological Incubators, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Thermo Scientific™ Heratherm™ Advanced Protocol Security Microbiological Incubators, which include a unique 140 °C decontamination routine, lockable incubator door with alarm for restricted access and automatic over- and under-temperature alarms.
- Advanced digital timer for daily or weekly ON / OFF cycles
- Easy to clean, stainless-steel interior (AISI 304)
- Intuitive user interface for setting temperature
- Dual convection for application versatility (except for large capacity units)—fan speed adjustable from 0 to 100%
- Optional sample sensor provides exact temperature of sample on display
- Microprocessor control with vacuum fluorescent display
It offers robust and corrosion-resistant surface that meets the demanding needs of pharmaceutical and clinical laboratories.
Certifications: Certified by an accredited microbiological institute.
Spécifications
- Catalog No:
- 10124-600
- 10124-616
- 10124-598
- 10124-612
- 10124-610
- 10124-606
- Temperature range:
- Ambient +5…105 °C
- Ambient +5…105 °C
- Ambient +5…105 °C
- Ambient +5…105 °C
- Ambient +5…105 °C
- Ambient +5…105 °C
- Temperature uniformity:
- ± 0.6 °C (fan off)
- ± 0.6 °C (fan off)
- ± 0.6 °C (fan off)
- ± 0.6 °C (fan off)
- ± 0.6 °C (fan off)
- ± 0.6 °C (fan off)
- Electrical:
- 120V, 60 Hz
- 120V, 60 Hz
- 120V, 60 Hz
- 120V, 60 Hz
- 120V, 60 Hz
- 120V, 60 Hz