Ward's® Essentials Trigonometry Puzzle Set
# de catalogue 470229-064
Fournisseur: Wards
Spécifications
- Description:Ward's® Essentials Trigonometry Puzzle Set
- No. of Lab Groups:1
- Cat. no.:470229-064
Spécifications
A propos de cet article
Relate the angles and sides in a right triangle in a hands on way.
- Durable materials for years of use
- A concrete demonstration of a difficult mathematical concept
Students will determine the functions associated with a right triangle using a precut series of right trianles. High quality precision-cut Birch verner pure bond plywood, Contains: Step-by-step educational guide, Includes the unit circle, three 30-60-90 triangles, two 45-45-90 triangles, two circular sectors, Kepler's triangle.
Customers Who Bought This Also Bought