Chemware® PTFE Balls, SAINT GOBAIN PERFORMANCE PLASTIC
Fournisseur: Saint Gobain Performance Plastics
Certificats
A propos de cet article
Chemware® PTFE balls are mainly used in column packing and mixed beds
- Minimize “bumping” during boiling
- Resistant to most chemicals
- Non-contaminating
They are inert and can be used up to 550 °F (288 °C).
Packaging: 100 balls per package.
Customers Who Bought This Also Bought