PTFE Valves, Two-Way and Three-Way, Burkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Two-way/Three-way valves are made of solid PTFE with hose nozzles
- Resistant to compression; 1 bar (14.50 psi) maximum
- Vacuum suitable up to 10-1 mmHg
- Autoclavable
They are resistant to chemicals, and offer very easy connection due to excellent anti-friction properties. Blue handles are made of polypropylene.
Customers Who Bought This Also Bought