CONSTIX®, Sealed Polyester Swabs, SP-6 Series, Contec®
Sealed Polyester Swabs, SP-6 Series, 148 mm (5.8"), 3.2 mm (0.13"), 15.8 mm (0.62"), 6.9 mm (0.27"), 163.8 mm (6.4")
Supplier: CONTEC, INC. MX
Fournisseur #: SP-6
CONSTIX®, Sealed Polyester Swabs, SP-6 Series, Contec®
Catalog # 76057-730
Supplier: CONTEC, INC. MX
Jump to:
Product Details & Documents
CONSTIX® SP-6 sealed polyester swabs consist of 100% knit polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
- Flexible tip provides a softer application on surfaces
- Polyester head provides enhanced sorbency, particle entrapment, and removal
- 100% polyester headis compatible with most common solvents, such as alcohols and ketones
The swab head is made of cleanroom laundered, double-layer knitted polyester. The polyester head is constructed without adhesives or binders. The swab is durable, absorbent, and features extremely low particles and residue levels.
Spécifications
- Désignation:Sealed Polyester Swabs, SP-6 Series
- Handle Length:148 mm (5.8")
- Overall Length:163.8 mm (6.4")
- Head Width:6.9 mm (0.27")
- Head Length:15.8 mm (0.62")
- Handle Width:3.2 mm (0.13")