A propos de cet article
When the situation calls for high performance, these neoprene gloves provide superior resistance to chemicals, punctures, and abrasions with a secure grip for handling small parts and slippery objects.
- Pure neoprene prevents latex allergic reactions
- Textured surface delivers excellent grip
- Ergonomic design for comfort
- Optimal thickness for durability and tactile sensitivity
- Powder-free to prevent contamination
- Tear-resistant beaded cuff eases donning
- Tested for use with chemotherapy drugs
- SHA Dermatologically Approved
The SW difference: 100% air inspected, 75% less residue, consistent tip to cuff.
Born of SW® manufacturing capabilities and advancements in material science, PowerChem™ offers chemical protection for highly specific applications. Using the most advanced materials and SW®'s proven manufacturing techniques, PowerChem™ provides the optimum blend of tactility and protection in a 5 mil glove. Ergonomically designed for comfort, this 1.5 AQL glove features a fully textured surface for secure grip in a variety of wet and dry environments and is approved for use with chemotherapy gloves.
Tested for use with chemotherapy drugs, PowerChem™ is also Dermatologically Approved by the Skin Health Alliance and has met stringent standards that ensure skin safety.
Biodegradable in landfills: 92.6% biodegradable in 2.5 years. ASTM D5526-12 test results do not predict future biodegradation. EcoTek® provides all the benefits of biodegradability without sacrificing performance, safety, or durability.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76051-188
- 76051-224
- 76051-156
- 76051-222
- 77463-352
- Model:
- PC-95GR
- PC-95GR
- PC-95GR
- PC-95GR
- PC-95GR
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Chloroprene
- Chloroprene
- Chloroprene
- Chloroprene
- Chloroprene
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes