SW® PowerForm® PF-95BL Blue 6.1 mil Nitrile Exam Dry Core® Sweat Absorbent Gloves
Supplier: Avantor
Certificates
About this item
Keep hands cooler and more comfortable with premium quality nitrile gloves, featuring Unique Dry Core™ moisture management technology.
- Dry Core® moisture management technology pulls sweat away from the skin leaving the wearer dryer
- Fully textured surface for superior wet and oily grip
- Silicone-free for critical environments
- 1.5 AQL exceeds laboratory standards
- ANSI level 3 abrasion
- EcoTek® sustainable technology
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; SHA Dermatologically Approved; Meets stringent standards that ensure skin safety; EAC TP TC 019/2011; CE 0493; CE EN 455; ANSI Level 3 Abrasion.
Ordering information: Glove sizes are as follows; S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Specifications
- Catalog No:
- 76051-000
- 76051-008
- 76051-006
- 76051-004
- 76051-002
- Model:
- PF-95BL
- PF-95BL
- PF-95BL
- PF-95BL
- PF-95BL
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- DriTek Hydrex Absorbent
- DriTek Hydrex Absorbent
- DriTek Hydrex Absorbent
- DriTek Hydrex Absorbent
- DriTek Hydrex Absorbent
- Finish:
- Fully Textured
- Fully Textured
- Fully Textured
- Fully Textured
- Fully Textured
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff length:
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- 242 mm (9.5")
- Cuff thickness:
- 5.1 mil (0.13 mm)
- 5.1 mil (0.13 mm)
- 5.1 mil (0.13 mm)
- 5.1 mil (0.13 mm)
- 5.1 mil (0.13 mm)
- Finger thickness:
- 6.9 mil (0.18 mm)
- 6.9 mil (0.18 mm)
- 6.9 mil (0.18 mm)
- 6.9 mil (0.18 mm)
- 6.9 mil (0.18 mm)
- Palm thickness:
- 6.1 mil (0.16 mm)
- 6.1 mil (0.16 mm)
- 6.1 mil (0.16 mm)
- 6.1 mil (0.16 mm)
- 6.1 mil (0.16 mm)
- FDA regulated:
- +
- +
- +
- +
- +
- Tensile strength:
- 20 MPa
- 20 MPa
- 20 MPa
- 20 MPa
- 20 MPa
- Ultimate elongation:
- 560%
- 560%
- 560%
- 560%
- 560%
- EN norm:
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- Food contact approved:
- +
- +
- +
- +
- +
- ASTM No.:
- D6319-19; F1671-13; F739-12
- D6319-19; F1671-13; F739-12
- D6319-19; F1671-13; F739-12
- D6319-19; F1671-13; F739-12
- D6319-19; F1671-13; F739-12