SP Wilmad-LabGlass Heavy-Duty Glass Stirrer Shafts, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Made of borosilicate glass, these heavy-duty, precision ground, polished stirrer shafts have a 19 mm diameter and are equipped with a side button for attaching stirrer blades (LG-9515 or LG-9516T).
The rod length below contact surface of bearing is unground and varies with flask capacity.
Customers Who Bought This Also Bought