VWR® Reusable Mohr-Style, Color-Coded, TD Pipettes, Class B
Supplier: VWR International
Certificates
About this item
VWR® Mohr pipettes are made from Borosilicate glass, ASTM 438, Type 1, Class B standard.
- Designed as straight glass tubes
- With one tapering end
- They are calibrated into small divisions
- Various amounts of liquid can be measured within the same device
- Mohr pipettes are calibrated to deliver a precise volume at a precise temperature
- They are used for accurate measurement and delivery of liquid volumes
The VWR® Mohr Pipettes Calibration and toleration standards for these pipettes are defined as per standard specifications and practices: ASTM E1044, ASTM E1293, ASTM E542, and ASTM E694.
Frequently Bought Together