VWR® Graduated Addition Funnels with PTFE Stopcocks
Fournisseur: VWR International
Certificats
A propos de cet article
VWR® Graduated Addition/Dropping funnels with PTFE Stopcocks are made from Borosilicate 3.3 glass, ASTM 438, Type 1, Class A standard.
Calibrated 'to Contain', flasks are graduated with a double metric, white ceramic enamel scale.
Customers Who Bought This Also Bought