A propos de cet article
For the quantification of 11-keto testosterone in plasma and other sample matrices.
- Detect the primary androgenic steroid in fish
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure 11-keto testosterone levels down to 1.3 pg/mL
- Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
- Mid-point: 50% B/B0: 2 – 10 pg/mL
11-KT is a second key androgenic steroid found in fish. It occurs in males together with testosterone in amounts which vary from less than 1 ng/mL to as much as 50 – 100 ng/mL, depending on the species and the stage of the reproductive cycle. Cayman's 11-keto Testosterone ELISA Kit is a competitive assay tha can be used for quantification of 11-keto testosterone in plasma and other sample matrices.