VWR® Buret with Straight Bore Glass Stopcock, Class B
# de catalogue 75997-922
Fournisseur: VWR International
UL Listed
Spécifications
- Capacity:50 ml
- O.D.:13 mm
- Tolerance:±0.10 ml
- Subdivisions:0.1 mL
Spécifications
A propos de cet article
VWR® Class B Buret made from 3.3 expansion Borosilicate glass
- Burets are supplied with a chemically-resistant, self-lubricating PTFE stopcock plug
- Used primarily for titration, to deliver one reactant until the precise endpoint is reached
- Calibration and tolerances are defined in the standard specifications and practices: ASTM E287, ASTM E542, ASTM E694, and ISO 385
The graduations are typically calibrated "To Deliver" (marked "TD" or "EX") at 20 to 27°C and indicated correspondingly on a label.
These Burettes are calibrated to deliver a precise volume at a particular temperature.
Customers Who Bought This Also Bought