Hydro Electric Power Station Demonstration
# de catalogue 470230-190
Fournisseur: Wards
Spécifications
- Description:Hydro Electric Power Station Demonstration
- Cat. no.:470230-190
Spécifications
A propos de cet article
Model demonstrates conversation of mechanical energy into electrical energy.
- Demonstrate conversion of mechanical energy into electrical energy
- Transparent cover allows you to see the turbine working
- Directly connected to a dynamo to generate electricity
This model demonstrates the conversion of mechanical energy into electrical energy, specifically a water turbine. Turbine is fitted with a transparent cover to show inner workings with inlet and outlet tubes. This is directly connected to a dynamo to generate electricity.
Customers Who Bought This Also Bought