ContactUsLinkComponent
Equal Mass Density Set
Equal Mass Density Set
# de catalogue 470230-180
Fournisseur:  Wards
Equal Mass Density Set
Equal Mass Density Set
# de catalogue 470230-180
Fournisseur:  Wards
Spécifications
  • Description:
    Equal Mass Density Set
  • Cat. no.:
    470230-180
Spécifications
A propos de cet article
Cylinder set for demonstrating density investigation and specific heat experiments.
  • All cylinders have same diameter to show principles of mass and density between various materials
  • Acrylic, nylon, PVC, aluminum, and brass cylinders
Equal mass density cylinder set of different materials. 5 cylinders included: Acrylic, Nylon, PVC, Aluminum, and Brass.
Customers Who Bought This Also Bought