ContactUsLinkComponent
Vibration Generator
Vibration Generator
# de catalogue 470230-260
Fournisseur:  Wards
Vibration Generator
Vibration Generator
# de catalogue 470230-260
Fournisseur:  Wards
Spécifications
  • Description:
    Vibration Generator
  • Cat. no.:
    470230-260
Spécifications
A propos de cet article
Provides electronically controlled waves.
  • Product is used for generating waves with a string or for use with a ripple tank
  • Overall height: 5.5", overall diameter: 4.5"
  • Plastic item with foam inside
  • For use with square wave generator
The vibration generator is a sturdy, user-friendly tool for providing electronically controlled waves - this allows one to create ripples for the study of wave motion (recommended for use with a square wave generator).
Customers Who Bought This Also Bought