Organ Pipe
# de catalogue 470230-256
Fournisseur: Wards
Spécifications
- Description:Organ Pipe
- Cat. no.:470230-256
Spécifications
A propos de cet article
Polished wood organ pipe for investigating beat phenomenon.
- Polished wooden organ pipe
- Fully extends to 34.5"
- Marked with chromatic scale from C(512) to (1024)
- Ideal for investigating beat phenomenon using a second pipe
Labs polished wooden organ pipe with movable piston is marked with the chromatic scale from C(512) to (1024). It has a fully extended length of 34.5" and works well for investigating the phenomenon of beats in conjunction with a second pipe of the same type.
Customers Who Bought This Also Bought