Sympathetic Tuning Fork on Wooden Base
# de catalogue 470230-252
Fournisseur: Wards
Spécifications
- Description:Sympathetic Tuning Fork on Wooden Base
- Cat. no.:470230-252
Spécifications
A propos de cet article
Steel tuning forks for physics experiments.
- Used to hear dissonance
- One fork adjustable
- Comes with mallet and bases
For demonstrating the production of beat frequencies. Comprises two nickel plated forks A (440 Hz ±5%) mounted on individual boxes. One fork is provided with adjustable mass by means of which frequency may be shifted from the nominal 440 Hz .When both forks are sounded a clearly audible 'beat' is produced, its rate depending upon the difference in frequency between the forks. (2) Tuning forks with bases, (1) Mallet.
Note: These are designed for physics experiments to create beats and are often not exactly 440 Hz (usually 436 - 444 Hz). They can be adjusted to create beats within each other or to match each other.
Customers Who Bought This Also Bought