Aluminum Tuning Fork, Set with Hammer
# de catalogue 470230-250
Fournisseur: Wards
Spécifications
- Description:Aluminum Tuning Fork, Set with Hammer
- Cat. no.:470230-250
Spécifications
A propos de cet article
Aluminum tuning forks for physics experiments.
- C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512)
- Comes with mallet and wood case
- Designed for physics experiments and actual frequencies can be ±5% stated Hz
Premium aluminum physics tuning fork set with hammer. Tuning forks included: C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512). Designed for Physics experiments and actual frequencies can be ±5% stated Hz.
Customers Who Bought This Also Bought