Steel Tuning Forks, Set of 8
# de catalogue 470230-246
Fournisseur: Wards
Spécifications
- Description:Steel Tuning Forks, Set of 8
- Cat. no.:470230-246
Spécifications
A propos de cet article
Steel tuning forks for physics experiments.
- ±5% of stated frequency
Steel Tuning Forks, Set of 8, C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512).
Customers Who Bought This Also Bought