Steel Tuning Forks with Wood Case
# de catalogue 470230-244
Fournisseur: Wards
Spécifications
- Description:Steel Tuning Forks with Wood case
- Cat. no.:470230-244
Spécifications
A propos de cet article
Steel tuning forks for physics experiments.
- Supplied in wooden case
Nickel-plated, plain shanks, with frequency marked, C (256), D (288), E (320), F (341), G (384), A (426), B (480), C (512). Set supplied in wooden case.
Customers Who Bought This Also Bought