Wave Apparatus, Plastic
Wave Apparatus, Plastic
Catalog # 470230-242
Supplier: Wards
Fournisseur #: PH0712N8
Jump to:
Product Details & Documents
Plastic wave apparatus ideal for studying wave behavior.
- Demonstrate longitudinal and transverse waves
This advanced apparatus demonstrates both longitudinal and transverse wave motions. Consisting of a number of eccentric discs supporting a series of metal rods revolving on a handle, transverse waves are obtained. Longitudinal waves are obtained from the bent rods.
Spécifications
- Désignation:Wave Apparatus, Plastic