Thermocouple Copper Constantan
# de catalogue 470230-176
Fournisseur: Wards
Spécifications
- Description:Thermocouple Copper Constantan
- Cat. no.:470230-176
Spécifications
A propos de cet article
Simple Thermocouple for demonstrating wide range of temperatures.
- Type T thermocouple
- Copper and Constantan
- Great repeatability at lower temperatures
- Well suited to oxidizing atmospheres
Very simple T-Type thermocouple comprised of copper and constantan. Use electricity to measure temperature. Great for demonstrations in the lab or classroom or for simply measuring a wide range of temperatures.
Customers Who Bought This Also Bought