Static Electricity Accessories
# de catalogue 470230-322
Fournisseur: Wards
Spécifications
- Description:Static Electricity Accessories
- Cat. no.:470230-322
Spécifications
A propos de cet article
Accessories for electrostatic generators.
Put your electrostatic generator (van de Graaff or Wimshurst) to use to do more than create static charge. Physically demonstrate numerous effects with this kit.
Ordering information: Kit includes (1) Dancing Ball Cylinder, (1) Plume of Hair with connector, (1) Plastic Comb, (1) Point Source Needle, (1) Neon Light Indicator, (1) Spinning Spiked Arm Wheel and (1) Pith ball on String and Cup.
Customers Who Bought This Also Bought