Convection of Gas Apparatus, Small
# de catalogue 470230-170
Fournisseur: Wards
Spécifications
- Description:Convection of Gas Apparatus, Small
- Cat. no.:470230-170
Spécifications
A propos de cet article
Apparatus to study convection of gas.
- Smaller sized to fit in any classroom than standard convection of gas apparatus
- Candles not included
This apparatus is comprised of a metal box with a sliding front door, two glass chimneys and a candle holder. Candles are not included.
Customers Who Bought This Also Bought